Learn More
Biolegend Brilliant Violet 421™ anti-ERK1/2 Phospho (Thr202/Tyr204)
Supplier:  Biolegend 369510
Encompass_Preferred
Brilliant Violet 421™ anti-ERK1/2 Phospho (Thr202/Tyr204) [6B8B69]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. 50-113-0202