Learn More
Biolegend Brilliant Violet 421™ anti-human IgM
Supplier:  Biolegend 314516
Encompass_Preferred
Brilliant Violet 421™ anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1127573