Learn More
Biolegend Brilliant Violet 421™ Rat IgG1, λ Isotype Ctrl
Supplier:  Biolegend 401911
Encompass_Preferred
Brilliant Violet 421™ Rat IgG1, λ Isotype Ctrl [G0114F7]; Isotype: Rat IgG1, λ; Reactivity: Rat; Apps: FC; Size: 50 ug
Catalog No. 50-712-226