Learn More
Biolegend Brilliant Violet 421™ Rat IgG1, κ isotype Ctrl
Supplier:  Biolegend 400429
Encompass_Preferred
Brilliant Violet 421™ Rat IgG1, κ isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Apps: FC, ICFC; Size: 125 uL
Catalog No. 50-168-940