Learn More
Biolegend Brilliant Violet 421™ Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400535
Encompass_Preferred
Brilliant Violet 421™ Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 125 uL
Catalog No. 50-168-949