Learn More
Biolegend Brilliant Violet 421™ Streptavidin
Supplier:  Biolegend 405225
Encompass_Preferred
Brilliant Violet 421™ Streptavidin Reactivity: Human, Mouse, Rat, All Species; Apps: FC, ICFC, ICC, IHC-F, IHC-P; Size: 100 ug
Catalog No. 50-169-086