Learn More
Biolegend Brilliant Violet 421™ anti-mouse/human CD45R/B220
Supplier:  Biolegend 103251
Encompass_Preferred
Brilliant Violet 421™ anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC, IHC-F; Size: 50 ug
Catalog No. 50-604-831