Learn More
Biolegend Brilliant Violet 510™ anti-human CD4
Supplier:  Biolegend 300546
Encompass_Preferred
Brilliant Violet 510™ anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-711-957