Learn More
Biolegend Brilliant Violet 510™ anti-human IgD
Supplier:  Biolegend 348219
Encompass_Preferred
Brilliant Violet 510™ anti-human IgD [IA6-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-6857