Learn More
Biolegend Brilliant Violet 510™ anti-human FcεRIα
Supplier:  Biolegend 334626
Encompass_Preferred
Brilliant Violet 510™ anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6183