Learn More
Biolegend Brilliant Violet 510™ anti-mouse CD90.2
Supplier:  Biolegend 105335
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1112002