Learn More
Biolegend Brilliant Violet 510™ anti-mouse CD14
Supplier:  Biolegend 123323
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD14 [Sa14-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-0505