Learn More
Biolegend Brilliant Violet 510™ anti-mouse CD226 (DNAM-1)
Supplier:  Biolegend 133607
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD226 (DNAM-1) [TX42.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-207-0973