Learn More
Biolegend Brilliant Violet 510™ anti-mouse CD86
Supplier:  Biolegend 105039
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-605-958