Learn More
Biolegend Brilliant Violet 510™ anti-mouse TCR β chain
Supplier:  Biolegend 109233
Encompass_Preferred
Brilliant Violet 510™ anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F; Size: 125 uL
Catalog No. 50-605-972