Learn More
Biolegend Brilliant Violet 510™ anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116237
Encompass_Preferred
Brilliant Violet 510™ anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-403-002