Learn More
Biolegend Brilliant Violet 510™ Mouse IgG2a, κ isotype Ctrl
Supplier:  Biolegend 400267
Encompass_Preferred
Brilliant Violet 510™ Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC, ICFC; Size: 25 tests
Catalog No. 50-403-551