Learn More
Biolegend Brilliant Violet 510™ Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400346
Encompass_Preferred
Brilliant Violet 510™ Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-604-697