Learn More
Biolegend Brilliant Violet 510™ anti-human CD56 (NCAM)
Supplier:  Biolegend 318340
Encompass_Preferred
Brilliant Violet 510™ anti-human CD56 (NCAM) [HCD56]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-604-139