Learn More
Biolegend Brilliant Violet 570™ anti-human CD45
Supplier:  Biolegend 304033
Encompass_Preferred
Brilliant Violet 570™ anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-166-071