Learn More
Biolegend Brilliant Violet 570™ anti-mouse CD19
Supplier:  Biolegend 115535
Encompass_Preferred
Brilliant Violet 570™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-163-561