Learn More
Biolegend Brilliant Violet 570™ anti-mouse CD8a
Supplier:  Biolegend 100739
Encompass_Preferred
Brilliant Violet 570™ anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-162-501