Learn More
Apexbio Technology LLC BV6 1001600-56-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062839
Encompass_Preferred
BV6 (CAS 1001600-56-1) is a selective inhibitor targeting the inhibitor of apoptosis protein (IAP) family The IAP family includes proteins such as XIAP c-IAP1 and c-IAP2 that regulate programmed cell death pathways BV6 induces apoptosis and sensitizes cancer cells to chemotherapeutic agents and radiation by decreasing expression levels of IAPs including XIAP and c-IAP1 In vitro treatment with BV6 exhibited an IC50 of 7 2 M in H460 non-small cell lung carcinoma cells and enhanced their response to radiation BV6 is employed in research exploring cancer cell apoptosis modulation and therapeutic resistance
Catalog No. 50-000-62839 Supplier Apexbio Technology LLC Supplier No. B46535.1
BV6 (CAS 1001600-56-1) is a selective inhibitor targeting the inhibitor of apoptosis protein (IAP) family The IAP family includes proteins such as XIAP c-IAP1 and c-IAP2 that regulate programmed cell death pathways BV6 induces apoptosis and sensitizes cancer cells to chemotherapeutic agents and radiation by decreasing expression levels of IAPs including XIAP and c-IAP1 In vitro treatment with BV6 exhibited an IC50 of 7 2 M in H460 non-small cell lung carcinoma cells and enhanced their response to radiation BV6 is employed in research exploring cancer cell apoptosis modulation and therapeutic resistance