Learn More
Biolegend Brilliant Violet 605™ anti-human CD127 (IL-7Rα)
Supplier:  Biolegend 351333
Encompass_Preferred
Brilliant Violet 605™ anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-604-353