Learn More
Biolegend Brilliant Violet 605™ anti-human CD194 (CCR4)
Supplier:  Biolegend 359418
Encompass_Preferred
Brilliant Violet 605™ anti-human CD194 (CCR4) [L291H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-604-632