Learn More
Biolegend Brilliant Violet 605™ anti-human CD24
Supplier:  Biolegend 311124
Encompass_Preferred
Brilliant Violet 605™ anti-human CD24 [ML5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee ; Apps: FC; Size: 100 tests
Catalog No. 50-207-3486