Learn More
Biolegend Brilliant Violet 605™ anti-human CD326 (Ep-CAM)
Supplier:  Biolegend 324224
Encompass_Preferred
Brilliant Violet 605™ anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-604-174