Learn More
Biolegend Brilliant Violet 605™ anti-human CD40
Supplier:  Biolegend 334336
Encompass_Preferred
Brilliant Violet 605™ anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey ; Apps: FC; Size: 100 tests
Catalog No. 50-850-661