Learn More
Biolegend Brilliant Violet 605™ anti-human Ki-67
Supplier:  Biolegend 350522
Encompass_Preferred
Brilliant Violet 605™ anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC; Size: 100 tests
Catalog No. 50-850-723