Learn More
Biolegend Brilliant Violet 605™ anti-human TIGIT (VSTM3)
Supplier:  Biolegend 372711
Encompass_Preferred
Brilliant Violet 605™ anti-human TIGIT (VSTM3) [A15153G]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1287259