Learn More
Biolegend Brilliant Violet 605™ anti-human CD16
Supplier:  Biolegend 360727
Encompass_Preferred
Brilliant Violet 605™ anti-human CD16 [B73.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7939