Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD11c
Supplier:  Biolegend 117334
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-604-920