Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD150 (SLAM)
Supplier:  Biolegend 115927
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD150 (SLAM) [TC15-12F12.2]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-402-996