Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD31
Supplier:  Biolegend 102427
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-850-267