Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD45.1
Supplier:  Biolegend 110738
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-604-896