Learn More
Biolegend Brilliant Violet 605™ anti-mouse IFN-γ
Supplier:  Biolegend 505840
Encompass_Preferred
Brilliant Violet 605™ anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 ug
Catalog No. 50-208-5709