Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD127 (IL-7Rα)
Supplier:  Biolegend 135025
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD127 (IL-7Rα) [A7R34]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-605-987