Learn More
Biolegend Brilliant Violet 605™ Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400161
Encompass_Preferred
Brilliant Violet 605™ Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 25 tests
Catalog No. 50-403-540