Learn More
Biolegend Brilliant Violet 605™ anti-mouse CX3CR1
Supplier:  Biolegend 149027
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CX3CR1 [SA011F11]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1502017