Learn More
Biolegend Brilliant Violet 650™ anti-human CD117 (c-kit)
Supplier:  Biolegend 313221
Encompass_Preferred
Brilliant Violet 650™ anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: FC; Size: 25 tests
Catalog No. 50-207-3602