Learn More
Biolegend Brilliant Violet 650™ anti-human CD183 (CXCR3)
Supplier:  Biolegend 353730
Encompass_Preferred
Brilliant Violet 650™ anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-103-0783