Learn More
Biolegend Brilliant Violet 650™ anti-human CD20
Supplier:  Biolegend 302336
Encompass_Preferred
Brilliant Violet 650™ anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-850-499