Learn More
Biolegend Brilliant Violet 650™ anti-human CD42b
Supplier:  Biolegend 303925
Encompass_Preferred
Brilliant Violet 650™ anti-human CD42b [HIP1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-207-3027