Learn More
Biolegend Brilliant Violet 650™ anti-human CD45RA
Supplier:  Biolegend 304135
Encompass_Preferred
Brilliant Violet 650™ anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-207-3051