Learn More
Biolegend Brilliant Violet 650™ anti-mouse/human Ki-67
Supplier:  Biolegend 151215
Encompass_Preferred
Brilliant Violet 650™ anti-mouse/human Ki-67 [11F6]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 50 ug
Catalog No. 50-207-2018