Learn More
Biolegend Brilliant Violet 650™ anti-mouse TCR β chain
Supplier:  Biolegend 109251
Encompass_Preferred
Brilliant Violet 650™ anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1675933