Learn More
Biolegend Brilliant Violet 650™ anti-mouse CD206 (MMR)
Supplier:  Biolegend 141723
Encompass_Preferred
Brilliant Violet 650™ anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 50 ug
Catalog No. 50-605-062