Learn More
Biolegend Brilliant Violet 650™ anti-mouse CD80
Supplier:  Biolegend 104731
Encompass_Preferred
Brilliant Violet 650™ anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 125 uL
Catalog No. 50-402-969