Learn More
Biolegend Brilliant Violet 650™ anti-mouse F4/80
Supplier:  Biolegend 123149
Encompass_Preferred
Brilliant Violet 650™ anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-103-0543