Learn More
Biolegend Brilliant Violet 650™ anti-mouse IgD
Supplier:  Biolegend 405721
Encompass_Preferred
Brilliant Violet 650™ anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-605-437